Benstat™ Divider Tote Boxes, Metro™
Product Details & Documents
Tote boxes offer safe and efficient storage of items. Boxes are durable, stackable, and feature high impact strength. There is a smooth area on each side of the box that is a convenient place to affix labels and bar codes.
Insert style covers fit inside the rim of the tote box and feature a finger hole that enables easy removal. Card holders are also available. Choose from holders made from Benstat™ or vinyl.
Insert style covers fit inside the rim of the tote box and feature a finger hole that enables easy removal. Card holders are also available. Choose from holders made from Benstat™ or vinyl.
Boxes are durable, stackable, and feature high impact strength. There is a smooth area on each side of the box that is a convenient place to affix labels and bar codes.
Documents
TDS Metro® Divider Tote Box Line
Datasheet
Especificaciones
- Capacity:2.8 L (0.10 cu.ft.)
- Interior Dimensions:16.5W×23.2L×8.6H cm (61/2×91/8×33/8")
- Exterior Dimensions:21W×27.6L×8.9H cm (81/4×107/8×31/2")
- Max. No. of Dividers:5 long, 7 short



