Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

8" Test Sieves, W.S. Tyler™

Stainless Frame-Stainless Cloth, 600 µm, #30
Catalog # 57332-077
Supplier:   W.S. Tyler
 :  5367
8" Test Sieves, W.S. Tyler™
Catalog # 57332-077
Supplier:  W.S. Tyler
8" Test Sieves, W.S. Tyler™
8" Test Sieves, W.S. Tyler™

Stainless Frame-Stainless Cloth, 600 µm, #30

Price
$4,350.53
Log in Para disponibilidad
Each (1UN)
Product Details & Documents

Overall Height - Full: 25/8"; Half: 15/8"

Especificaciones
  • Description:
    Stainless Frame-Stainless Cloth
  • Mesh Size:
    600 µm
  • USA Series:
    #30