Orders LinkContactUsLinkComponent
Nunc® CryoColor Coder Inserts, Thermo Scientific
Green
  24270-106
 :  355018
Nunc® CryoColor Coder Inserts, Thermo Scientific
  24270-106
Nunc® CryoColor Coder Inserts, Thermo Scientific
Nunc® CryoColor Coder Inserts, Thermo Scientific
Green
$2,190.28
Paquete de 500 Case de 4 (500UN)
For comprehensive sample identification, fits inside CryoTube® vial cap.
  • Made of polystyrene
  • Available in 10 different colors
 : Assortment 24270-120 includes either 50 of each color (for a pack) or 200 of each color (for a case).
  • Color:
    Green