STERI-Buffer®, Veltek Associates
Phosphate buffer solution, 90 mL (3.0 oz.)
Supplier: VELTEK ASSOCIATES, INC. MX
: SB100-90
STERI-Buffer®, Veltek Associates
Catalog # 14222-664
Supplier: VELTEK ASSOCIATES, INC. MX
CAS Number:
Jump to:
Product Details & Documents
Developed for use as a buffer solution and diluent in lab testing such a bioburden testing, pharmaceutical microbial limits testing, and AET testing and as a rinsing agent."
Sealed with a “no tamper” strip. Filtered at 0.2µm. Gamma-irradiated, double-bagged, and delivered with lot-specific sterile documentation.
Documents
Certificates
Buscar por número de loteEspecificaciones
Especificaciones
- Description:Phosphate buffer solution
- Size:90 mL (3.0 oz.)



