Orders LinkContactUsLinkComponent
Technical Buffer Solutions, Sachets
±0.02, Sachet, red, 30×20 ml, 4.01
  11238-438
 :  51302069
Technical Buffer Solutions, Sachets
  11238-438
 :  
Technical Buffer Solutions, Sachets
Technical Buffer Solutions, Sachets
±0.02, Sachet, red, 30×20 ml, 4.01
 :  
$4,618.17
Paquete de 30
NIST-traceable buffer for pH electrode calibration and verification.
  • NIST-traceable
  • Ensure freshness
  • Improve data
Also available in multiple packaging options.
Mettler Toledo buffer sachets ensure a fresh portion of buffer is used for each calibration, improving your pH data.
  • Pack type:
    Sachet, red
  • Size:
    30×20 ml
  • pH:
    4.01
  • pH Accuracy:
    ±0.02
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 