Viscometer Holders, Cannon
Cannon-Manning Semi-Micro Removeable Viscometer Holder, Plastic
47735-278
: 9726-M50
All CANNON viscometer holders fit holes 51 mm (2" ) in diameter.
- Self-aligning
- Constructed of chrome-plated machined brass
- All metal holders are provided with a handle
Plastic holders are self-aligning and hold the viscometer firmly by means of a spring clip. Rubber holders are made of neoprene rubber.
- Description:Cannon-Manning Semi-Micro Removeable Viscometer Holder
- Material:Plastic