Order Entry
Mexico
Orders LinkContactUsLinkComponent
Hydroxylammonium chloride, ReagentPlus® analytical reagent

Hydroxylammonium chloride, ReagentPlus® analytical reagent

Plastic bottle, wide mouth, 100 g
Catalog # 220039-048
Corrosivo
Salud
Irritante
Medio ambiente
Número de proveedor #:  159417-100G
Hydroxylammonium chloride, ReagentPlus® analytical reagent
Catalog # 220039-048
CAS Number:  5470-11-1
Hydroxylammonium chloride, ReagentPlus® analytical reagent

Plastic bottle, wide mouth, 100 g

CAS Number:  5470-11-1
Price
$1,621.80
Log in Para disponibilidad
Each (100g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Sinónimos:  Hydroxylamine HCl, Hydroxylamine hydrochloride
  • CAS Number:5470-11-1
  • Fórmula:NH₂OH·HCl
  • Número MDL:MFCD00051089
  • Molecular Weight:69.49 g/mol
  • Índice Merck:13,04853
  • Punto de ebullición:306 °C (1013 hPa)
  • Punto de fusión:151 °C
  • Densidad:1.67 g/cm³ (20 °C)
  • Temperatura de almacenamiento:Ambient
  • EINECS:226-798-2
  • UN:2923
  • ADR:8,III

Documents

Especificaciones

Especificaciones

  • Pack type:
    Plastic bottle, wide mouth
  • Size:
    100 g

Especificaciones

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.