$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
TAPA DE CIERRE 18MM AC INOX
89209-368
:
CHEMGLASS INC.
TAPA DE CIERRE 18MM AC INOX
89209-368
:
CHEMGLASS INC.
:
CLS-1502-018
Frequently Bought Together
VWR® Disposable Serological Pipettes
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Universal Pipette Tips
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH