Orders LinkContactUsLinkComponent
Bare Copper Wire
Bare Copper Wire
Bare copper wire, 16 AWG
Catalog # 470206-062
Supplier #:  B16
Bare Copper Wire
Catalog # 470206-062
Bare Copper Wire
Bare copper wire, 16 AWG
Price
$225.48
Log in Para disponibilidad
Each (4oz)
Product Details & Documents
Bare copper wire, #16, ¼ lb.
Especificaciones
  • Description:
    Bare copper wire
  • Gauge:
    16 AWG