Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
QUICKTASK CABEZAL AZUL PP 40CM
Catalog # 89201-650
Supplier:
CONTEC, INC. MX
Certificates
Expand image
QUICKTASK CABEZAL AZUL PP 40CM
Catalog # 89201-650
Supplier:
CONTEC, INC. MX
Supplier Number:
2740
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
89201-650_694865-0-0-1
89201-650_744352-0-0-1
89201-650_744354-0-0-1
89201-650_796005-0-0-1
89201-650_807530-0-0-1
Product Literature
Contec Mopping Products for Critical Environments Brochure v2
Documentation
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International