Orders LinkContactUsLinkComponent
BDH® pH Test Strips
0.3/0.4, 6.0 - 7.7
  BDH35317.604
 :   BDH
BDH® pH Test Strips
  BDH35317.604
 :  BDH
BDH® pH Test Strips
BDH® pH Test Strips
0.3/0.4, 6.0 - 7.7
$432.06
Each (1UN) Paquete de 5 (1UN) Case de 40 (1UN)
Used for rapid pH testing directly at the point of interest, strips are always ready for immediate use. They are also suitable for careful testing of dangerous, poisonous, or aggressive liquids as the long handle prevents fingers from coming into contact with the sample. The dye is chemically bound to the test strip and cannot bleed into the sample, protecting against contamination and enabling measurement in weakly buffered or strongly alkaline solutions. Included color chart has 4 different color blocks for each pH value, enabling highly precise pH determination. Strips are available in 13 different pH ranges with accuracy down to 0.2 pH units. Dimensions: 85Lx6Wmm.
  • Designed for safe testing of dangerous, poisonous, or aggressive liquids
  • Long handle protects user from contact with sample
  • Fixed dye cannot bleed into the sample, protecting it against contamination and enabling measurements even in weakly buffered or strongly alkaline solutions
Dimensions: (LxW) 85x6 mm.
 : Strips are packaged 100 to a box. Included color chart has 4 different color blocks for each pH value
  • pH Range:
    6.0 - 7.7
  • pH Graduation:
    0.3/0.4
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.