Orders LinkContactUsLinkComponent
VWR® Glove Liners
VWR® Glove Liners
Half Finger Liners, with Green Cuff, S, White
Catalog # 94000-616
Supplier:   VWR International
VWR® Glove Liners
Catalog # 94000-616
Supplier:  VWR International
VWR® Glove Liners
VWR® Glove Liners
Half Finger Liners, with Green Cuff, S, White
Price
$654.36
Log in Para disponibilidad
Paquete de 20 Case de 15 (20UN)
Product Details & Documents
Ergonomically designed glove liners wick away moisture for comfortable use with disposable gloves.
  • Ambidextrous glove liners
  • Reduces perspiration and wicks away moisture
  • Low-lint, 100% 15 denier nylon composition
  • Sturdy, launderable, and reusable
  • Color-coded cuffs indicate size
  • Pre-laundered; will not shrink
Liners provide protection and comfort when used with latex, nitrile, and PVC gloves. Fully launderable for reusability and economy.
Packaging: Glove Liners are packaged 20 pair per bag, 15 bags per case. They are packed in clean polyethylene bags that are free of silicone, dioctyl phthalate (DOP), and amides.
Especificaciones
  • Glove Color:
    White
  • Glove size:
    S
  • Description:
    Half Finger Liners, with Green Cuff
  • Cat. No.:
    94000-616
  • Disposable / reusable:
    Reusable
  • Cuff style:
    Knit wrist
  • Ambidextrous / hand-specific:
    Ambidextrous