VWR® Glove Liners
Full Finger Liners, with Green Cuff, S, White
Supplier: VWR International
VWR® Glove Liners
Catalog # 94000-608
Supplier: VWR International
Product available on GSA Advantage®
Jump to:
Product Details & Documents
Ergonomically designed glove liners wick away moisture for comfortable use with disposable gloves.
- Ambidextrous glove liners
- Reduces perspiration and wicks away moisture
- Low-lint, 100% 15 denier nylon composition
- Sturdy, launderable, and reusable
- Color-coded cuffs indicate size
- Pre-laundered; will not shrink
Liners provide protection and comfort when used with latex, nitrile, and PVC gloves. Fully launderable for reusability and economy.
Packaging: Glove Liners are packaged 20 pair per bag, 15 bags per case. They are packed in clean polyethylene bags that are free of silicone, dioctyl phthalate (DOP), and amides.
Documents
Especificaciones
- Glove Color:White
- Glove size:S
- Description:Full Finger Liners, with Green Cuff
- Cat. No.:94000-608
- Disposable / reusable:Reusable
- Cuff style:Knit wrist
- Ambidextrous / hand-specific:Ambidextrous