Orders LinkContactUsLinkComponent
Universal Pipet Tips, Neptune™
Universal Pipet Tips, Neptune™
Universal Fit Tips, 200 µl, Yellow, Bulk
Catalog # 89140-108
Supplier:   Avantor
Universal Pipet Tips, Neptune™
Catalog # 89140-108
Supplier:  Avantor
Universal Pipet Tips, Neptune™
Universal Pipet Tips, Neptune™
Universal Fit Tips, 200 µl, Yellow, Bulk
Price
$341.97
Log in for availability
Paquete de 1,000 Case de 10 (1,000,UN)
Product Details & Documents
These universal pipet tips are made from virgin polypropylene. The thin wall design ensures an optimal seal and easy ejection with industry leading pipettors. Eco-friendly racks are made from recycled polypropylene and feature a unique compact packaging configuration that reduces the carbon footprint.
  • Autoclavable
  • Good sustainability
  • Superior stability, firm placement
  • Maximum space saving design
Eco-friendly empty pipette tips trays are manufactured from recycled polypropylene. When combined with ESP™ reload systems, these tip trays reduce plastic waste by up to 90%.
The thin wall design ensures an optimal seal and easy ejection with industry leading pipettors. Eco-friendly racks are made from recycled polypropylene and feature a unique compact packaging configuration that reduces the carbon footprint.
Certifications: Certified free of RNase, DNase, pyrogens, and PCR Inhibitors. Manufactured under stringent controls in an ISO 9001 certified facility. Each and every lot is tested for accuracy and precision, ensuring superior quality and compatibility. All raw materials are controlled.
Ordering information: Bulk orders are packaged 1000 tips per bag. Rack & Stack packaging contains five inserts of 192 tips.
Specifications
  • Volume:
    200 µl
  • Tip Color:
    Yellow
  • Packed:
    Bulk
  • Tip Style:
    Universal Fit Tips