Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

PYREX® Hydrometer Cylinders, Plain, Corning

500 ml, 50 mm, 390 mm
  89090-762
Product available on GSA Advantage®
   Low Manufacturing Impact
 :   Corning
 :  2962-500
PYREX® Hydrometer Cylinders, Plain, Corning
  89090-762
 :  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
PYREX® Hydrometer Cylinders, Plain, Corning
PYREX® Hydrometer Cylinders, Plain, Corning

500 ml, 50 mm, 390 mm

 
$1,180.00
   
Paquete de 1 Case de 12 (1UN)
 

These PYREX® glas hydrometer cylinders have a large hexagonal base sealed to the cylinder body to increase stability. Breakage is minimized by the heavy wall construction.

  • Excellent chemical resistance
  • Good temperature resistance

Meets ASTM specifications D-287 and E-100.

 
  • Ø:
    50 mm
  • Capacity:
    500 ml
  • Height:
    390 mm
  • Cat. No.:
    89090-762
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Environmentally Preferable:
    Low Manufacturing Impact