Orders LinkContactUsLinkComponent
PYREX® Crystallizing Dishes, Corning
190×100 mm
  89090-700
 :   Corning
 :  3140-190
PYREX® Crystallizing Dishes, Corning
  89090-700
 :  Corning
PYREX® Crystallizing Dishes, Corning
PYREX® Crystallizing Dishes, Corning
190×100 mm
$5,370.00
Paquete de 2 Case de 3 (2UN)
PYREX® borosilicate glass, without spout.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Flat bottom and straight sides. Rims reinforced and fire-polished to reduce chipping.
Documents
  • Diameter ×H:
    190×100 mm