PYREX® Crystallizing Dishes, Corning
170×90 mm
Supplier: Corning
Número de proveedor #: 3140-170
Jump to:
Product Details & Documents
PYREX® borosilicate glass, without spout.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Flat bottom and straight sides. Rims reinforced and fire-polished to reduce chipping.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Diameter ×H:170×90 mm