Orders LinkContactUsLinkComponent
PYREX® Crystallizing Dishes, Corning
PYREX® Crystallizing Dishes, Corning
170×90 mm
Catalog # 89090-698
Supplier:   Corning
Número de proveedor #:  3140-170
PYREX® Crystallizing Dishes, Corning
Catalog # 89090-698
Supplier:  Corning
PYREX® Crystallizing Dishes, Corning
PYREX® Crystallizing Dishes, Corning
170×90 mm
Price
$4,540.00
Log in Para disponibilidad
Paquete de 2 Case de 4 (2UN)
Product Details & Documents
PYREX® borosilicate glass, without spout.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Flat bottom and straight sides. Rims reinforced and fire-polished to reduce chipping.
Documents
Especificaciones
  • Diameter ×H:
    170×90 mm