Orders LinkContactUsLinkComponent
VWR® Spatulas, Porcelain
VWR® Spatulas, Porcelain
25.4 cm (10"), 6 ml
Catalog # 89038-210
Supplier:   VWR International
VWR® Spatulas, Porcelain
Catalog # 89038-210
Supplier:  VWR International
VWR® Spatulas, Porcelain
VWR® Spatulas, Porcelain
25.4 cm (10"), 6 ml
Price
$477.27
Log in Para disponibilidad
Each (1UN) Case de 8 (1UN) Paleta de 1,272 (1UN)
Product Details & Documents
Chemical-porcelain spatulas feature a spatula on one end and a spoon on the other.
  • High chemical resistance
  • Chemical-porcelain construction
  • Glazed for added protection
Especificaciones
  • Capacity:
    6 ml
  • Length:
    25.4 cm (10")