Orders LinkContactUsLinkComponent
VWR® Hirsch Filter Funnels
10 ml, 10 mm, 42 mm
  89038-136
VWR® Hirsch Filter Funnels
  89038-136
VWR® Hirsch Filter Funnels
VWR® Hirsch Filter Funnels
10 ml, 10 mm, 42 mm
$182.18
Each (1UN) Case de 12 (1UN) Paleta de 3,360 (1UN)
These chemical-porcelain funnels feature a fixed perforated plate and glazing inside and out, except for rim.
  • Highly resistant to chemicals
  • Autoclavable
Filter funnels are available in a range of volume capacities and top outside diameters.
  • Capacity:
    10 ml
  • Top O.D.:
    42 mm
  • Fits Paper Diameter:
    10 mm