Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Buchner Filter Funnels

VWR® Buchner Filter Funnels

1860 ml, 270 mm, 185 mm
Catalog # 89038-132
Supplier:   VWR International
VWR® Buchner Filter Funnels
Catalog # 89038-132
Supplier:  VWR International
VWR® Buchner Filter Funnels
VWR® Buchner Filter Funnels

1860 ml, 270 mm, 185 mm

Price
$1,709.08
Log in Para disponibilidad
Each (1UN) Case de 2 (1UN)
Product Details & Documents

These chemical-porcelain Buchner filter funnels feature a fixed perforated plate and glazing inside and out, except for the rim.

  • Highly resistant to chemicals
  • Autoclavable
  • Glazed inside and out, except for the rim
Especificaciones
  • Capacity:
    1860 ml
  • Height:
    270 mm
  • Fits Paper Diameter:
    185 mm