Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Buchner Filter Funnels

VWR® Buchner Filter Funnels

1060 ml, 221 mm, 142–150 mm
Catalog # 89038-130
Supplier:   VWR International
VWR® Buchner Filter Funnels
Catalog # 89038-130
Supplier:  VWR International
VWR® Buchner Filter Funnels
VWR® Buchner Filter Funnels

1060 ml, 221 mm, 142–150 mm

Price
$976.53
Log in Para disponibilidad
Each (1UN) Case de 2 (1UN)
Product Details & Documents

These chemical-porcelain Buchner filter funnels feature a fixed perforated plate and glazing inside and out, except for the rim.

  • Highly resistant to chemicals
  • Autoclavable
  • Glazed inside and out, except for the rim
Especificaciones
  • Capacity:
    1060 ml
  • Height:
    221 mm
  • Fits Paper Diameter:
    142–150 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.