Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Buchner Filter Funnels

VWR® Buchner Filter Funnels

550 ml, 195 mm, 110 mm
Catalog # 89038-126
Supplier:   VWR International
VWR® Buchner Filter Funnels
Catalog # 89038-126
Supplier:  VWR International
VWR® Buchner Filter Funnels
VWR® Buchner Filter Funnels

550 ml, 195 mm, 110 mm

Price
$527.23
Log in Para disponibilidad
Each (1UN) Case de 4 (1UN)
Product Details & Documents

These chemical-porcelain Buchner filter funnels feature a fixed perforated plate and glazing inside and out, except for the rim.

  • Highly resistant to chemicals
  • Autoclavable
  • Glazed inside and out, except for the rim
Especificaciones
  • Capacity:
    550 ml
  • Height:
    195 mm
  • Fits Paper Diameter:
    110 mm