Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Buchner Filter Funnels

VWR® Buchner Filter Funnels

30 ml, 89 mm, 35–40 mm
Catalog # 89038-118
Supplier:   VWR International
VWR® Buchner Filter Funnels
Catalog # 89038-118
Supplier:  VWR International
VWR® Buchner Filter Funnels
VWR® Buchner Filter Funnels

30 ml, 89 mm, 35–40 mm

Price
$208.54
Log in Para disponibilidad
Each (1UN) Case de 12 (1UN) Paleta de 2,112 (1UN)
Product Details & Documents

These chemical-porcelain Buchner filter funnels feature a fixed perforated plate and glazing inside and out, except for the rim.

  • Highly resistant to chemicals
  • Autoclavable
  • Glazed inside and out, except for the rim
Especificaciones
  • Capacity:
    30 ml
  • Height:
    89 mm
  • Fits Paper Diameter:
    35–40 mm