VWR® Buchner Filter Funnels
2 ml, 64 mm, 13 mm
Catalog # 89038-116
Supplier: VWR International
Jump to:
Product Details & Documents
These chemical-porcelain Buchner filter funnels feature a fixed perforated plate and glazing inside and out, except for the rim.
- Highly resistant to chemicals
- Autoclavable
- Glazed inside and out, except for the rim
Documents
Especificaciones
- Capacity:2 ml
- Height:64 mm
- Fits Paper Diameter:13 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



