Orders LinkContactUsLinkComponent
KIMAX® Solid Borosilicate Glass Beads, Kimble Chase
KIMAX® Solid Borosilicate Glass Beads, Kimble Chase
5 mm, 3000/container; 125/cu.in.
Catalog # 89001-518
Número de proveedor #:  13500-5
KIMAX® Solid Borosilicate Glass Beads, Kimble Chase
Catalog # 89001-518
KIMAX® Solid Borosilicate Glass Beads, Kimble Chase
KIMAX® Solid Borosilicate Glass Beads, Kimble Chase
5 mm, 3000/container; 125/cu.in.
Price
$5,089.01
Log in Para disponibilidad
Case (1UN)
Product Details & Documents
Highly resistant to attack by most cold liquids, hot liquids, and vapors
Suitable for use as mixing beads, boiling stones, or packing for distillation columns. Beads are durable and will not disintegrate or affect delicate compounds.
Ordering information: Packed in 0.45kg (1lb.) containers with a total volume of 360cm3 (22 cu. in.).
Especificaciones
  • Ø:
    5 mm
  • No. of Beads:
    3000/container; 125/cu.in.