KIMAX® Solid Borosilicate Glass Beads, Kimble Chase
4 mm, 5200/container; 250/cu.in.
Catalog # 89001-516
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Número de proveedor #: 13500-4
KIMAX® Solid Borosilicate Glass Beads, Kimble Chase
Catalog # 89001-516
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Jump to:
Product Details & Documents
Highly resistant to attack by most cold liquids, hot liquids, and vapors
Suitable for use as mixing beads, boiling stones, or packing for distillation columns. Beads are durable and will not disintegrate or affect delicate compounds.
Ordering information: Packed in 0.45kg (1lb.) containers with a total volume of 360cm3 (22 cu. in.).
Documents
Especificaciones
- Ø:4 mm
- No. of Beads:5200/container; 250/cu.in.



