Orders LinkContactUsLinkComponent
KIMAX® Round-Bottom Boiling Flasks, Kimble Chase
KIMAX® Round-Bottom Boiling Flasks, Kimble Chase
3 L, 24/40, 185 x 255 mm
Catalog # 89001-294
KIMAX® Round-Bottom Boiling Flasks, Kimble Chase
Catalog # 89001-294
Supplier:  
KIMAX® Round-Bottom Boiling Flasks, Kimble Chase
KIMAX® Round-Bottom Boiling Flasks, Kimble Chase
3 L, 24/40, 185 x 255 mm
Discontinued
El producto ha sido descontinuado. Sin embargo, encontramos productos alternativos similares.
Alternativas
FLASKRND BTTM S-NK CS1 3000ML Catalog #29126-103
Product 29126-103 is similar to 89001-294 but manufacturer and specifications may vary.
Product Details & Documents
Short neck flasks are designed to ASTM Specification E1403, Class 3 requirements
They feature a full-length, outer, [ST] 24/40 joint.
Documents
Especificaciones
  • ST Joint No.:
    24/40
  • Capacity:
    3 L
  • O.D.×H:
    185 x 255 mm