KIMAX® Round-Bottom Boiling Flasks, Kimble Chase
3 L, 24/40, 185 x 255 mm
Catalog # 89001-294
Jump to:
Discontinued
El producto ha sido descontinuado. Sin embargo, encontramos productos alternativos similares.
Alternativas
FLASKRND BTTM S-NK CS1 3000ML Catalog #29126-103
Product 29126-103 is similar to 89001-294 but manufacturer and specifications may vary.
Product Details & Documents
Short neck flasks are designed to ASTM Specification E1403, Class 3 requirements
They feature a full-length, outer, [ST] 24/40 joint.
Documents
Certificates
Buscar por número de loteKimble Petrochemical Glassware Brochure
Documentación del producto
Especificaciones
- ST Joint No.:24/40
- Capacity:3 L
- O.D.×H:185 x 255 mm