Orders LinkContactUsLinkComponent
KIMAX® Flat-Bottom Boiling Flasks, Kimble Chase
KIMAX® Flat-Bottom Boiling Flasks, Kimble Chase
1 L, 24/40, 130 x 182 mm
Catalog # 89001-268
Número de proveedor #:  25055-1000
KIMAX® Flat-Bottom Boiling Flasks, Kimble Chase
Catalog # 89001-268
KIMAX® Flat-Bottom Boiling Flasks, Kimble Chase
KIMAX® Flat-Bottom Boiling Flasks, Kimble Chase
1 L, 24/40, 130 x 182 mm
Price
$1,003.65
Log in Para disponibilidad
Paquete de 1 Case de 12 (1UN)
Product Details & Documents
All flasks have a short neck and a full length [ST] 24/40 interchangeable ground glass joint
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Documents
Especificaciones
  • ST Joint No.:
    24/40
  • Capacity:
    1 L
  • O.D.×H:
    130 x 182 mm