KIMAX® Flat-Bottom Boiling Flasks, Kimble Chase
500 ml, 24/40, 102 x 155 mm
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Número de proveedor #: 25055-500
KIMAX® Flat-Bottom Boiling Flasks, Kimble Chase
Catalog # 89001-266
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Jump to:
Product Details & Documents
All flasks have a short neck and a full length [ST] 24/40 interchangeable ground glass joint
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Documents
Certificates
Buscar por número de loteKimble Petrochemical Glassware Brochure
Documentación del producto
Especificaciones
- ST Joint No.:24/40
- Capacity:500 ml
- O.D.×H:102 x 155 mm