KIMAX® Flat-Bottom Boiling Flasks, Kimble Chase
125 ml, 24/40, 65 x 115 mm
Catalog # 89001-262
: 25055-125
Jump to:
Discontinued
El producto ha sido descontinuado. Sin embargo, encontramos productos alternativos similares.
Alternativas
Product Details & Documents
All flasks have a short neck and a full length [ST] 24/40 interchangeable ground glass joint
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Especificaciones
- ST Joint No.:24/40
- Capacity:125 ml
- O.D.×H:65 x 115 mm