Orders LinkContactUsLinkComponent
KIMAX® Flat-Bottom Boiling Flasks, Kimble Chase
125 ml, 24/40, 65 x 115 mm
Catalog # 89001-262
 :  25055-125
KIMAX® Flat-Bottom Boiling Flasks, Kimble Chase
Catalog # 89001-262
Supplier:  
KIMAX® Flat-Bottom Boiling Flasks, Kimble Chase
KIMAX® Flat-Bottom Boiling Flasks, Kimble Chase
125 ml, 24/40, 65 x 115 mm
Discontinued
El producto ha sido descontinuado. Sin embargo, encontramos productos alternativos similares.
Alternativas
FLASK SHRT NECK PYREX 125 ML Catalog #89091-136
Product Details & Documents
All flasks have a short neck and a full length [ST] 24/40 interchangeable ground glass joint
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Especificaciones
  • ST Joint No.:
    24/40
  • Capacity:
    125 ml
  • O.D.×H:
    65 x 115 mm