KIMAX® Solid Borosilicate Glass Beads, Kimble Chase
3 mm, 12000/container; 550/cu.in.
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
: 13500-3
KIMAX® Solid Borosilicate Glass Beads, Kimble Chase
Catalog # 89001-052
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Jump to:
Product Details & Documents
Highly resistant to attack by most cold liquids, hot liquids, and vapors
Suitable for use as mixing beads, boiling stones, or packing for distillation columns. Beads are durable and will not disintegrate or affect delicate compounds.
: Packed in 0.45kg (1lb.) containers with a total volume of 360cm3 (22 cu. in.).
Especificaciones
- Ø:3 mm
- No. of Beads:12000/container; 550/cu.in.