Orders LinkContactUsLinkComponent
VITLAB® Measuring Scoops, HDPE, BrandTech
VITLAB® Measuring Scoops, HDPE, BrandTech
350 ml (11.8 oz.), 31 cm (12¼")
Catalog # 83008-708
Número de proveedor #:  40593
VITLAB® Measuring Scoops, HDPE, BrandTech
Catalog # 83008-708
VITLAB® Measuring Scoops, HDPE, BrandTech
VITLAB® Measuring Scoops, HDPE, BrandTech
350 ml (11.8 oz.), 31 cm (12¼")
Price
$737.27
Log in Para disponibilidad
Paquete de 6
Product Details & Documents
One-piece, molded measuring scoops made of high density polyethylene.
  • Eight different sizes
  • High quality
VITLAB® measuring scoops are precision molded from durable high-density polyethylene. Handles are cotoured, with a tapered filling edge. Scoops are food-safe according to EC Directive No. 10/2011.
Especificaciones
  • Capacity:
    350 ml (11.8 oz.)
  • Length:
    31 cm (12¼")