Orders LinkContactUsLinkComponent
VITLAB® Measuring Scoops, HDPE, BrandTech
150 ml (5 oz.), 25 cm (9⅞")
  83008-706
 :  40493
VITLAB® Measuring Scoops, HDPE, BrandTech
  83008-706
VITLAB® Measuring Scoops, HDPE, BrandTech
VITLAB® Measuring Scoops, HDPE, BrandTech
150 ml (5 oz.), 25 cm (9⅞")
$763.00
Paquete de 12
One-piece, molded measuring scoops made of high density polyethylene.
  • Eight different sizes
  • High quality
VITLAB® measuring scoops are precision molded from durable high-density polyethylene. Handles are cotoured, with a tapered filling edge. Scoops are food-safe according to EC Directive No. 10/2011.
  • Capacity:
    150 ml (5 oz.)
  • Length:
    25 cm (9⅞")