VITLAB® Measuring Scoops, HDPE, BrandTech
150 ml (5 oz.), 25 cm (9⅞")
83008-706
: 40493
One-piece, molded measuring scoops made of high density polyethylene.
- Eight different sizes
- High quality
VITLAB® measuring scoops are precision molded from durable high-density polyethylene. Handles are cotoured, with a tapered filling edge. Scoops are food-safe according to EC Directive No. 10/2011.
- Capacity:150 ml (5 oz.)
- Length:25 cm (9⅞")