VWR® Specimen Forceps, Serrated
Curved, 203 mm (8")
Supplier: VWR International
Jump to:
Product Details & Documents
Specimen forceps are ergonomically designed for safely and efficiently removing specimens from deep jars or bottles.
- Stainless steel construction
- Serrated tips
- Easy to use
Durable specimen forceps are built to withstand years of regular use.
Documents
Especificaciones
- Description:Curved
- Length:203 mm (8")