Orders LinkContactUsLinkComponent
VWR® Specimen Forceps, Serrated
VWR® Specimen Forceps, Serrated
Straight, 152 mm (6")
Catalog # 82027-438
Supplier:   VWR International
VWR® Specimen Forceps, Serrated
Catalog # 82027-438
Supplier:  VWR International
VWR® Specimen Forceps, Serrated
VWR® Specimen Forceps, Serrated
Straight, 152 mm (6")
Price
$171.10
Log in Para disponibilidad
Each (1UN) Paquete de 12 (1UN) Case de 60 (1UN)
Product Details & Documents
Specimen forceps are ergonomically designed for safely and efficiently removing specimens from deep jars or bottles.
  • Stainless steel construction
  • Serrated tips
  • Easy to use
Durable specimen forceps are built to withstand years of regular use.
Especificaciones
  • Description:
    Straight
  • Length:
    152 mm (6")