Orders LinkContactUsLinkComponent
VWR® Specimen Forceps
VWR® Specimen Forceps
305 mm (12")
Catalog # 82027-382
Supplier:   VWR International
VWR® Specimen Forceps
Catalog # 82027-382
Supplier:  VWR International
VWR® Specimen Forceps
VWR® Specimen Forceps
305 mm (12")
Price
$312.50
Log in Para disponibilidad
Each (1UN) Paquete de 12 (1UN) Case de 60 (1UN)
Product Details & Documents
Specimen forceps are ergonomically designed for safely and efficiently removing specimens from deep jars or bottles.
  • Stainless steel construction
  • Straight tips
  • Easy to use
Durable specimen forceps are built to withstand years of regular use.
Documents
Especificaciones
  • Length:
    305 mm (12")