Orders LinkContactUsLinkComponent
VWR® Microscope Slide Boxes for 50 Slides
Blue
  82024-580
VWR® Microscope Slide Boxes for 50 Slides
  82024-580
VWR® Microscope Slide Boxes for 50 Slides
VWR® Microscope Slide Boxes for 50 Slides
Blue
$158.51
Each (1UN) Paquete de 24 (1UN) Case de 72 (1UN)
Ideal for storage or transport, these durable ABS plastic boxes accommodate 50 slides apiece.
  • Durable ABS plastic outer shell
  • Color-coded for easy identification
  • Accepts all standard slides
Boxes feature cork lining and a secure clasp to ensure safety in transit or storage. All accept standard 25x75mm (1x3") slides. Boxes are color-coded for easy identification and their stackable design increases transit and storage convenience. Dimensions: 21x8.5x3.2 cm (85/16x311/32x11/4").
  • Color:
    Blue