VWR® Microscope Slide Boxes for 100 Slides
Yellow
Catalog # 82003-412
Supplier: VWR International
Jump to:
Product Details & Documents
Each box features cork lining and an outer shell made of durable ABS plastic.
- Durable ABS plastic outer shell
- Stores up to 100 standard (1x3") slides
Secured with a rust-resistant nickel-plated clasp and hinge pin. Each slot is numbered to correspond to the slide inventory sheet on the inside cover of the box. For storage of up to 100 standard 25x75 mm (1x3") slides. Boxes stack securely on top of each other. Color-coded for easy identification. Dimensions: LxWxD:22.2x17.1x3.3 cm (83/4x63/4x15/16").
Documents
Certificates
Buscar por número de loteEspecificaciones
- Color:Yellow