TLC Developing Chamber, Cylindrical, Chemglass
TLC developing chamber, 60/15 joint size, 50×100 mm plates
Catalog # 80066-466
Supplier: CHEMGLASS INC.
: CG-1181-03
Jump to:
Product Details & Documents
Cylindrical glass chamber for development of TLC plates from 25x75mm to 50x100mm
Chamber has a short length inner with a tool ball style handle. Chambers 80066-462 and 80066-464 have a hex base for added stability. Complete chamber is supplied with bottle and stopper.
Especificaciones
- Description:TLC developing chamber, 60/15 joint size
- For Use With:50×100 mm plates