Orders LinkContactUsLinkComponent
TLC Developing Chamber, Cylindrical, Chemglass
TLC developing chamber, 60/15 joint size, 50×100 mm plates
Catalog # 80066-466
Supplier:   CHEMGLASS INC.
 :  CG-1181-03
TLC Developing Chamber, Cylindrical, Chemglass
Catalog # 80066-466
Supplier:  CHEMGLASS INC.
TLC Developing Chamber, Cylindrical, Chemglass
TLC Developing Chamber, Cylindrical, Chemglass
TLC developing chamber, 60/15 joint size, 50×100 mm plates
Price
$6,190.00
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Cylindrical glass chamber for development of TLC plates from 25x75mm to 50x100mm
Chamber has a short length inner with a tool ball style handle. Chambers 80066-462 and 80066-464 have a hex base for added stability. Complete chamber is supplied with bottle and stopper.
Especificaciones
  • Description:
    TLC developing chamber, 60/15 joint size
  • For Use With:
    50×100 mm plates