Orders LinkContactUsLinkComponent
PYREX® Watch Glasses, Corning
PYREX® Watch Glasses, Corning
Watch glass, 150 mm (529/32")
Catalog # 66110-123
Supplier:   Corning
Número de proveedor #:  9985-150
PYREX® Watch Glasses, Corning
Catalog # 66110-123
Supplier:  Corning
PYREX® Watch Glasses, Corning
PYREX® Watch Glasses, Corning
Watch glass, 150 mm (529/32")
Price
$3,280.00
Log in Para disponibilidad
Paquete de 12 Case de 6 (12UN)
Product Details & Documents
PYREX® borosilicate glass.
  • High chemical durability
  • Prevents contamination of solutions
These watch glasses are molded to the same radius of curvature for convenient stacking. The heavy wall and uniform fire-polished edge provides the mechanical strength. The outside diameter cannot be controlled within close limits and a slight degree of out-of-round can be expected.
They are also used as covers for heated beakers.
 : One package has 12 watch glasses.
Documents
Especificaciones
  • Ø:
    150 mm (529/32")
  • Description:
    Watch glass