PYREX® Watch Glasses, Corning
Watch glass, 150 mm (529/32")
Supplier: Corning
Número de proveedor #: 9985-150
Jump to:
Product Details & Documents
PYREX® borosilicate glass.
- High chemical durability
- Prevents contamination of solutions
These watch glasses are molded to the same radius of curvature for convenient stacking. The heavy wall and uniform fire-polished edge provides the mechanical strength. The outside diameter cannot be controlled within close limits and a slight degree of out-of-round can be expected.
They are also used as covers for heated beakers.
: One package has 12 watch glasses.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Ø:150 mm (529/32")
- Description:Watch glass