PYREX® Watch Glasses, Corning
Watch glass, 100 mm (315/16")
Supplier: Corning
Supplier #: 9985-100
Jump to:
Product Details & Documents
PYREX® borosilicate glass.
- High chemical durability
- Prevents contamination of solutions
These watch glasses are molded to the same radius of curvature for convenient stacking. The heavy wall and uniform fire-polished edge provides the mechanical strength. The outside diameter cannot be controlled within close limits and a slight degree of out-of-round can be expected.
They are also used as covers for heated beakers.
Packaging: One package has 12 watch glasses.
Documents
Certificates
Search by Lot NumberSpecifications
- Ø:100 mm (315/16")
- Description:Watch glass