Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

VWR® Bacti Cell Spreaders

Yellow, 24.0 cm, 6.0 cm, Sterile
Catalog # 60828-688
Supplier:   VWR International
VWR® Bacti Cell Spreaders
Catalog # 60828-688
Supplier:  VWR International
VWR® Bacti Cell Spreaders
VWR® Bacti Cell Spreaders

Yellow, 24.0 cm, 6.0 cm, Sterile

Price
$2,361.97
Log in Para disponibilidad
Paquete de 25 Case de 4 (25UN)
Product Details & Documents

Convenient and economical cell spreaders are designed to simplify bacterial culture work.

  • Autoclavable
  • Simplify bacterial culture work
  • Durable fabrication

Durable enough to be autoclaved and reused.

 : Cell spreaders are available sterile (individually wrapped) or nonsterile (bulk packed).

Especificaciones
  • Sterility:
    Sterile
  • Color:
    Yellow
  • Blade Width:
    6.0 cm
  • Overall Length:
    24.0 cm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.