Orders LinkContactUsLinkComponent
Mantle/Controller Package, Series TM, Glas-Col®
250 ml, 180 W
  33752-409
 :   GLAS-COL
 :  100DTM102PL
Mantle/Controller Package, Series TM, Glas-Col®
  33752-409
 :  GLAS-COL
Mantle/Controller Package, Series TM, Glas-Col®
Mantle/Controller Package, Series TM, Glas-Col®
250 ml, 180 W
$22,729.94
Each (1UN)
Glas-Col® offers TM series mantles and PowrTrol power control (see 33797-414 series) together in a special package
CSA certified.
The compact control is a solid-state, proportional voltage manual control that provides precise control of the output range, adjustable from 3–100%. Control has a lighted on/off power switch, three-wire load receptacle, and three-wire cord with molded plug. Series TM mantles feature an aluminum exterior with fabric interior, units up to 600W, 120V, 5A. Mantles withstand an internal operating temperature of 450°C (840°F).
  • Power:
    180 W
  • For Flask Size:
    250 ml