VWR® Lead Ring Flask Weights
500 - 2000 ml, 56 mm (2.2"), 102 mm (4.0"), 700 g (1.5 lbs.)
Supplier: VWR International
Jump to:
Product Details & Documents
Protect and stabilize expensive glassware with Vikem® vinyl-coated lead rings.
- Stabilize glassware immersed in water baths
- Reduce floating, breakage and chemical spills
- Coated with bright red Vikem® vinyl
- Chemical-resistant
- Will not scratch flasks
- Available in five diameter/weight combinations
Vikem® vinyl coated lead rings stabilize glassware immersed in water baths, thereby reducing floating, breakage, and chemical spills.
Certificates
Buscar por número de loteEspecificaciones
- Fits Flask:500 - 2000 ml
- O.D.:102 mm (4.0")
- Weight:700 g (1.5 lbs.)
- I.D.:56 mm (2.2")