Orders LinkContactUsLinkComponent
IsoTip® Pipet Tips, Corning®
IsoTip® Pipet Tips, Corning®
1 - 200 µl, Most Research-Grade Pipettors, Extended Length, 96/Rack
Catalog # 29443-278
Supplier:   Corning
Número de proveedor #:  4810
IsoTip® Pipet Tips, Corning®
Catalog # 29443-278
Supplier:  Corning
IsoTip® Pipet Tips, Corning®
IsoTip® Pipet Tips, Corning®
1 - 200 µl, Most Research-Grade Pipettors, Extended Length, 96/Rack
Price
$3,090.00
Log in Para disponibilidad
Case (960UN)
Product Details & Documents
Designed for a variety of applications where elimination of aerosol contamination is critical, including Taq/PCR applications and radioisotope handling.
  • Exceptional durability
  • All Corning® tip racks stand up to the demands of use with multichannel pipettors and withstand repeated autoclaving cycles
  • Sterile and certified RNase-/ DNase-free and non pyrogenic
Especificaciones
  • Volume:
    1 - 200 µl
  • Packed:
    96/Rack
  • For Use With:
    Most Research-Grade Pipettors, Extended Length