IsoTip® Pipet Tips, Corning®
1 - 200 µl, Most Research-Grade Pipettors, Extended Length, 96/Rack
Supplier: Corning
Número de proveedor #: 4810
Jump to:
Product Details & Documents
Designed for a variety of applications where elimination of aerosol contamination is critical, including Taq/PCR applications and radioisotope handling.
- Exceptional durability
- All Corning® tip racks stand up to the demands of use with multichannel pipettors and withstand repeated autoclaving cycles
- Sterile and certified RNase-/ DNase-free and non pyrogenic
Especificaciones
- Volume:1 - 200 µl
- Packed:96/Rack
- For Use With:Most Research-Grade Pipettors, Extended Length