Order Entry
Mexico
Orders LinkContactUsLinkComponent
Nalgene® Erlenmeyer Flasks, Polypropylene, Thermo Scientific

Nalgene® Erlenmeyer Flasks, Polypropylene, Thermo Scientific

250 ml, 6
  29136-864
Supplier #:  4102-0250
Nalgene® Erlenmeyer Flasks, Polypropylene, Thermo Scientific
  29136-864
Nalgene® Erlenmeyer Flasks, Polypropylene, Thermo Scientific
Nalgene® Erlenmeyer Flasks, Polypropylene, Thermo Scientific

250 ml, 6

 
$2,710.00
   
Paquete de 6 Case de 2 (6UN)
 

Wide-mouth flasks are built with heavy walls for hard use and a wide base for stability.

  • Liquid level easily visible
  • Autoclavable

The 2L flask has molded-in graduations.

 
  • Capacity:
    250 ml
  • Stopper No.:
    6
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.