Upright Brooms, Perfex
Poly fiber brooms are ideal for sweeping in confined areas. Polypropylene fibers are fused onto a high-impact resistant polymer block. Flagged fibers are ideal for sweeping very fine and medium particles. Unflagged fibers are great for heavy and coarse materials, and for removing dirt and deposits from rough surfaces.
Polypropylene fibers are fused onto a high-impact resistant polymer block
Durable PVC handle (sold separately) locks securely in place with the self-locking Lite-N-Tite® mechanism. This handle/broom head connection system eliminates loosening and premature fiber fallout. Broom construction resists collecting contaminants. Excellent abrasion and chemical resistance.
Flagged fibers are ideal for sweeping very fine and medium particles. Unflagged fibers are great for heavy and coarse materials, and for removing dirt and deposits from rough surfaces.
: Broom heads and handle are ordered separately.
- Bristle Color:White
- Bristle Material:PP
- Bristle Trim:13.3 - 17.8 cm (5¼ - 7")
- Head Width:25.4L cm (10")