Orders LinkContactUsLinkComponent
VWR® Sharps Container Systems, Red
VWR® Sharps Container Systems, Red
1.6 gal., Non-Stackable, 25.4W×15.2D×27.9H cm (10W×6D×11"H), Red
Catalog # 19001-025
VWR® Sharps Container Systems, Red
Catalog # 19001-025
Supplier:  
VWR® Sharps Container Systems, Red
VWR® Sharps Container Systems, Red
1.6 gal., Non-Stackable, 25.4W×15.2D×27.9H cm (10W×6D×11"H), Red
Discontinued
Product Details & Documents
Sharps container systems provide solutions for the safe and effective containment and disposal of contaminated needles, syringes, and other sharps. Choose from three different systems in a variety of sizes.
  • Puncture-resistant
  • Autoclave-safe and incinerator-safe
  • Transparent, locking lids
  • Needle removal ports
The extended neck sharps containers feature a funneled vertical drop and extended handles. Puncture-resistant and autoclave- and incinerator-safe.
Specifications
  • Capacity:
    1.6 gal.
  • Color:
    Red
  • Dimensions:
    25.4W×15.2D×27.9H cm (10W×6D×11"H)
  • Style:
    Non-Stackable