VWR® Sharps Container Systems, Red
1.6 gal., Non-Stackable, 25.4W×15.2D×27.9H cm (10W×6D×11"H), Red
Jump to:
Discontinued
Product Details & Documents
Sharps container systems provide solutions for the safe and effective containment and disposal of contaminated needles, syringes, and other sharps. Choose from three different systems in a variety of sizes.
- Puncture-resistant
- Autoclave-safe and incinerator-safe
- Transparent, locking lids
- Needle removal ports
The extended neck sharps containers feature a funneled vertical drop and extended handles. Puncture-resistant and autoclave- and incinerator-safe.
Specifications
- Capacity:1.6 gal.
- Color:Red
- Dimensions:25.4W×15.2D×27.9H cm (10W×6D×11"H)
- Style:Non-Stackable