Orders LinkContactUsLinkComponent
VWR® Sharps Container Systems, Red
VWR® Sharps Container Systems, Red
1.7 qt., Non-Stackable, wth Needle Removal, 8.9W×8.9D×25.4H cm (3.5W×3.5D×10"H), Red
  19001-020
VWR® Sharps Container Systems, Red
  19001-020
 :  
VWR® Sharps Container Systems, Red
VWR® Sharps Container Systems, Red
1.7 qt., Non-Stackable, wth Needle Removal, 8.9W×8.9D×25.4H cm (3.5W×3.5D×10"H), Red
Sharps container systems provide solutions for the safe and effective containment and disposal of contaminated needles, syringes, and other sharps. Choose from three different systems in a variety of sizes.
  • Puncture-resistant
  • Autoclave-safe and incinerator-safe
  • Transparent, locking lids
  • Needle removal ports
The extended neck sharps containers feature a funneled vertical drop and extended handles. Puncture-resistant and autoclave- and incinerator-safe.
  • Capacity:
    1.7 qt.
  • Color:
    Red
  • Dimensions:
    8.9W×8.9D×25.4H cm (3.5W×3.5D×10"H)
  • Style:
    Non-Stackable, wth Needle Removal