VWR® Sharps Container Systems, Red
1.7 qt., Non-Stackable, 8.9W×8.9D×25.4H cm (3.5W×3.5D×10"H), Red
Catalog # 19001-016
Jump to:
Discontinued
Product Details & Documents
Sharps container systems provide solutions for the safe and effective containment and disposal of contaminated needles, syringes, and other sharps. Choose from three different systems in a variety of sizes.
- Puncture-resistant
- Autoclave-safe and incinerator-safe
- Transparent, locking lids
- Needle removal ports
The extended neck sharps containers feature a funneled vertical drop and extended handles. Puncture-resistant and autoclave- and incinerator-safe.
Especificaciones
- Capacity:1.7 qt.
- Color:Red
- Dimensions:8.9W×8.9D×25.4H cm (3.5W×3.5D×10"H)
- Style:Non-Stackable